Merck
20480
"Y" GlasSeal Connector, Borosilicate Glass
pkg of 1 ea
Informacje o tej pozycji
UNSPSC Code:
41121800
NACRES:
SB.54
Pomoc techniczna
Pozwól nam pomóc
packaging
pkg of 1 ea
technique(s)
gas chromatography (GC): suitable
General description
Quickly connect three pieces of fused silica tubing of the same or different diameters without the need for tools. The leak-free connection is useful to split a sample to two columns, or to split a column effluent to two detectors. Silanized for an inert inside surface. For use with 0.10 - 0.53 mm I.D. fused silica tubing.
Column connectors are useful for attaching a guard column/retention gap to an analytical column, or for repairing a broken column.
Other Notes
We offer a variety of chromatography accessories including analytical syringes
Legal Information
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)