Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
6 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 3150,00 zł |
Informacje o tej pozycji
3150,00 zł
material
spherical
sterility
sterile; autoclaved
feature
lid: no
manufacturer/tradename
MicroTissues Inc. 24-1
packaging
pack of (Contains 3 small and 3 large)
technique(s)
cell culture: suitable
application(s)
3D cell culture
General description
Since the agarose is transparent, the spheroids or microtissues that form at the bottom of each agarose micro-well can be easily viewed using a standard inverted microscope using phase contrast, bright field or fluorescence microscopy.
Features and Benefits
- The micro-molds are reusable up to 12 times and can be sterilized via a standard steam autoclave (30 min, dry cycle).
- The micro-molds should be stored in a covered container to maintain sterility and to avoid collecting dust or fibres on their small features.
- Any microscope with sufficient magnification and resolution functions for cell based views or images will be suitable for use with the 3D Petri Dish products.
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description 3 small and 3 large, fits 24 well plates | description 3 small and 3 large, fits 12 well plates | description size S, 8 x 12 array, fits 24 well plates | description size L, 9 x 9 array, fits 12 well plates |
| feature lid: no | feature lid: no | feature lid: no, 8 x 12 array | feature lid: no, 9 x 9 array |
| manufacturer/tradename MicroTissues Inc. 24-1 | manufacturer/tradename MicroTissues Inc. 12-1 | manufacturer/tradename MicroTissues Inc. 24-96 | manufacturer/tradename MicroTissues Inc. 12-81 |
| sterility sterile; autoclaved | sterility sterile; autoclaved | sterility sterile; autoclaved | sterility sterile; autoclaved |
| material spherical | material spherical | material spherical | material spherical |
| packaging pack of (Contains 3 small and 3 large) | packaging pack of (Contains 3 small and 3 large) | packaging pack of 6 ea | packaging pack of 6 ea |
| application(s) 3D cell culture | application(s) 3D cell culture | application(s) 3D cell culture | application(s) 3D cell culture |
Still not finding the right product?
Explore all of our products under MicroTissues® 3D Petri Dish® micro-mold mixed spheroid kit
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?