Informacje o tej pozycji
NACRES:
NB.43
UNSPSC Code:
41121804
eCl@ss:
32040501
Packaging:
pack of 1 ea
Pomoc techniczna
Pozwól nam pomócpackaging
pack of 1 ea
manufacturer/tradename
Worthington RS30-9C16
technique(s)
cryopreservation: suitable
General description
All Worthington cryostorage refrigerators and dewars each comes with a standard lid, so it is not necessary to purchase an additional one. However, many customers buy spare caps/necktube cores in order to modify them so that different kinds of sample storage devices can be suspended into a storage cannister.
Still not finding the right product?
Explore all of our products under Worthington Accessories
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?