Przejdź do zawartości
Z744002
Worthington Accessories
rack for 8 drawers for LS4800 liquid nitrogen refrigerator
Synonim(y):
366879, dewar draw rack, dewar holder, dewar rack, rack
Informacje o tej pozycji
NACRES:
NB.51
UNSPSC Code:
41122800
Manufacturer/tradename:
Worthington R05K-9C50
Packaging:
pack of 1 ea
Pomoc techniczna
Pozwól nam pomóc
packaging
pack of 1 ea
manufacturer/tradename
Worthington R05K-9C50
technique(s)
cryopreservation: suitable
General description
All Worthington cryostorage refrigerators and dewars each comes with a standard lid, so it is not necessary to purchase an additional one. However, many customers buy spare caps/necktube cores in order to modify them so that different kinds of sample storage devices can be suspended into a storage cannister.
Still not finding the right product?
Explore all of our products under Worthington Accessories
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)