Informacje o tej pozycji
NACRES:
NB.35
UNSPSC Code:
55121611
Manufacturer/tradename:
Diversified Biotech PSTK-1070
Packaging:
pack of 36 ea
Pomoc techniczna
Pozwól nam pomócpackaging
pack of 36 ea
manufacturer/tradename
Diversified Biotech PSTK-1070
General description
Quick Reference - Petristickers correspond to wells on a microplate for fast colony identification
High Contrast - Colored grid allows colonies to stand out from the Petri dish background
Updated Design - Rectangular shape leaves ample space above and below grid for writing on the dish
Two Layouts - Reference wells numerically (1-96) or alpha numerically (A1-H12)
Ideally Sized - PetriStickers™ are 2.8 in. x 1.75 in. (71 mm x 44.5 mm), to fit most common Petri dishes
High Contrast - Colored grid allows colonies to stand out from the Petri dish background
Updated Design - Rectangular shape leaves ample space above and below grid for writing on the dish
Two Layouts - Reference wells numerically (1-96) or alpha numerically (A1-H12)
Ideally Sized - PetriStickers™ are 2.8 in. x 1.75 in. (71 mm x 44.5 mm), to fit most common Petri dishes
Legal Information
PetriStickers is a trademark of Diversified Biotech
p3xFLAG is a trademark of Sigma-Aldrich Co. LLC
Still not finding the right product?
Explore all of our products under PetriStickers™
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?