Merck
Z743573
PetriStickers
6-sector pie format
Synonim(y):
Diversified Biotech PSTK-2000, dish sticker, laboratory labelling, petri dish stickers, sample identification label
Informacje o tej pozycji
NACRES:
NB.17
UNSPSC Code:
55121611
Manufacturer/tradename:
Diversified Biotech PSTK-2000
Packaging:
pack of 36 ea
Pomoc techniczna
Pozwól nam pomóc
packaging
pack of 36 ea
manufacturer/tradename
Diversified Biotech PSTK-2000
General description
Quick Reference - Petristickers correspond to wells on a microplate for fast colony identification
High Contrast - Colored grid allows colonies to stand out from the Petri dish background
Updated Design - Rectangular shape leaves ample space above and below grid for writing on the dish
Two Layouts - Reference wells numerically (1-96) or alpha numerically (A1-H12)
Ideally Sized - PetriStickers are 2.8 in. x 1.75 in. (71 mm x 44.5 mm), to fit most common Petri dishes
Legal Information
PetriStickers is a trademark of Diversified Biotech
p3xFLAG is a trademark of Sigma-Aldrich Co. LLC
Still not finding the right product?
Explore all of our products under PetriStickers
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)