Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 1780,00 zł |
Informacje o tej pozycji
1780,00 zł
packaging
pack of 1 ea
manufacturer/tradename
Benchmark Scientific BV1003-E-UK
General description
For the vortexing of single tubes, the cupped surface would generally be preferred. However, for applications requiring a flat/dimpled surface for mixing uniquely shaped vessels (or a handful of multiple test-tubes), the head can easily be flipped over and reattached in reverse orientation.
Like the original BenchMixer, the drive system is balanced with Q-Drive technology, providing a smooth vortexing action, while minimizing noise and excessive vibration. This results in quieter, more efficient and longer lasting operation. With a maximum speed up to 3,200 rpm, the mixer provides an instant vortex of even the largest sample sizes, including full 50ml tubes.
Features and Benefits
Powerful motor for INSTANT vortexing
Continuous or "touch" operation
Q-Drive dynamic balancing system
Variable speed from 200 to 3200 rpm
Wide variety of accessories, for tubes up to 50ml
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description AC/DC input 230 V AC, UK plug | description AC/DC input 230 V AC, Schuko plug | description AC/DC input 230 V AC, UK plug | description AC/DC input 230 V AC, European 2-pin plug, CE compliant |
| manufacturer/tradename Benchmark Scientific BV1003-E-UK | manufacturer/tradename Benchmark Scientific BV1003-E | manufacturer/tradename Benchmark Scientific BV1010-GB | manufacturer/tradename Benchmark Scientific BV1005-E with EU plug |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pkg of 1 ea | packaging - |
Benchmark, hybridization, micro sample tube, microsample, micro tube, mix, mixer, mixing, PCR, plate, sample tube, shake, shaker, shaking, vortex, vortexer | Benchmark, hybridization, micro sample tube, microsample, micro tube, mix, mixer, mixing, PCR, plate, sample tube, shake, shaker, shaking, vortex, vortexer | - | - |
Still not finding the right product?
Explore all of our products under BenchMixer™ V2 Vortexer with reversible top
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?