Przejdź do
Z742233
Labnet Fastpette Pro Pipette Controller Accessories
PTFE filter, autoclavable, pore size 0.2 μm, pack of 5
Synonim(y):
pipet
Informacje o tej pozycji
NACRES:
NB.01
UNSPSC Code:
41121500
Manufacturer/tradename:
Labnet P2023
Feature:
autoclavable
Material:
PTFE filter
Packaging:
pack of 5
material
PTFE filter
feature
autoclavable
packaging
pack of 5
manufacturer/tradename
Labnet P2023
pore size
0.2 μm
Legal Information
FastPette is a trademark of Labnet Intl., Inc.
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)