Merck
Z740419
Specimen containers with snap cap
size 1000 mL
Synonim(y):
Simport Plastics C580-5, sample container, sample storage, tamper proof sample storage
Informacje o tej pozycji
NACRES:
NB.17
UNSPSC Code:
41121800
Pomoc techniczna
Pozwól nam pomóc
material
polypropylene cap, polypropylene container
packaging
pack of 100 ea
manufacturer/tradename
Simport C580-5
size
1000 mL
General description
These disposable containers made of polypropylene, are ideal for collection and storage of fluids, powders, solids, pathology specimens and hazardous samples. Lids are tight fitting
Still not finding the right product?
Explore all of our products under Specimen containers with snap cap
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)