Przejdź do
Z740392
The SpecTainer™ I, tamper evident
size 120 mL, sterile
Synonim(y):
Simport Plastics C566-120CYS, sample receptacle, sample storage, sample tube, secure sample container
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
300 ea
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
3270,00 zł
Informacje o tej pozycji
NACRES:
NB.22
UNSPSC Code:
41121800
Manufacturer/tradename:
Simport C566-120CYS
Material:
polyethylene cap, polypropylene container
Packaging:
pack of 300 ea
3270,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
​
Pomoc techniczna
Pozwól nam pomóc
material
polyethylene cap, polypropylene container
sterility
sterile
packaging
pack of 300 ea
manufacturer/tradename
Simport C566-120CYS
size
120 mL
General description
For applications needing the utmost security where sample integrity is of high importance. A great feature of the Spectainer I is that it incorporates a unique tamper evident screw cap ensuring peace of mind during transport or storage situations where someone might have tampered with the specimen.
Both containers and caps are manufactured without the use of plasticisers or mold release agents. All material used is free from latex. Containers are 95kpa compliant. Available in three sizes: 60, 90 and 120 ml. The 60 ml size however is not graduated. The sterile container protects its sterility with a tamper evident seal.
Legal Information
SpecTainer is a trademark of Simport
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
Z740388Z740390Z740391
description
size 120 mL, sterile
description
size 60 mL, sterile
description
size 90 mL, sterile
description
size 90 mL, non-sterile
manufacturer/tradename
Simport C566-120CYS
manufacturer/tradename
Simport C566-60CYS
manufacturer/tradename
Simport C566-90CYS
manufacturer/tradename
Simport C566-90Y
material
polypropylene container, polyethylene cap
material
polyethylene cap, polypropylene container
material
polyethylene cap, polypropylene container
material
polyethylene cap, polypropylene container
packaging
pack of 300 ea
packaging
pack of 500 ea
packaging
pack of 400 ea
packaging
pack of 400 ea
size
120 mL
size
60 mL
size
90 mL
size
90 mL
sterility
sterile
sterility
sterile
sterility
sterile
sterility
non-sterile
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)