Merck
Z740237
TLC Plastic Sheets
L × W 20 cm × 20 cm, PEI-cellulose, matrix, binder, not applicable, fluorescent indicator
Informacje o tej pozycji
NACRES:
NC.07
UNSPSC Code:
41115711
Pomoc techniczna
Pozwól nam pomóc
Quality Segment
material
PEI-cellulose, matrix, plastic support
feature
binder not applicable, fluorescent indicator
packaging
pkg of 25 ea
manufacturer/tradename
EMD Millipore® 1.05579.0001
technique(s)
thin layer chromatography (TLC): suitable
L × W
20 cm × 20 cm
layer thickness
N/A
plate L × W
20 cm × 20 cm
particle size
10-12 μm
pore size
60 Å medium pore diameter
Legal Information
EMD Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Still not finding the right product?
Explore all of our products under TLC Plastic Sheets
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)