Merck
Z740161
LifeSep magnetic separator unit
384F, for 384 well tray
Synonim(y):
cell sorting magnetic seperation unit, Dexter Magnetic Technologies 2501007-1
Informacje o tej pozycji
NACRES:
NA.56
UNSPSC Code:
41100000
Pomoc techniczna
Pozwól nam pomóc
feature
for 384 well tray
manufacturer/tradename
Dexter Magnetic Technologies 2501007-1
technique(s)
cell culture: suitable
General description
Durable, easy-to-use and designed to accommodate industry standard microtitre plates. For use in cell sorting, RNA and DNA isolation and purification of biomolecules. Fast and easy aspiration is achieved with our Neodymium-Iron-Boron (Nd-Fe-B) magnet configuration.
Features and Benefits
  • Patented Nd-Fe-B magnet configuration for optimal results
  • Optimized field gradient within wells
  • Rapid separation of micro- and nanometer beads
  • Easy aspiration
  • Superior particle retention
  • Removable base for easy cleaning
Still not finding the right product?
Explore all of our products under LifeSep magnetic separator unit
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)