Przejdź do
Z726060
Spectra-Por® Float-A-Lyzer® G2
black, 1 mL, MWCO 3.5-5 kDa
Synonim(y):
Spectra-Por Float-A-Lyzer
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
Informacje o tej pozycji
NACRES:
NB.22
UNSPSC Code:
41123100
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomóc
material
black
packaging
pack of 12 ea (dry with glycerol)
manufacturer/tradename
Spectrum Labs G235029
technique(s)
dialysis: suitable
pore size
3.5-5 kDa MWCO
capacity
1 mL
General description
  • 95 - 98% Sample Recovery
  • Highest Membrane Purity
  • Superior Handling & Leak Prevention
  • Volume Specific Dilution Control
  • Complete & Ready-to-Use Packaging
Spectrum′s proprietary Biotech Grade CE is a low protein-binding synthetic membrane available in 4 precise MWCO′s (color coded for MWCO variants) with no heavy metal and sulfide contaminants. The cylindrical tubing geometry prevents sample dilution (associated with cassette-type devices) and provides open access for total volume retrieval. Only the Float-A-Lyzer® G2 assures a 95-98% sample recovery while maintaining 99% sample purity and <1% sample dilution.
The leak-proof screw-on cap with sealing o-ring provides easy access with included pipette for loading, in-process testing and sample retrieval, without the risk of needle punctures. The included floatation ring improves sample buoyancy and vertical orientation during dialysis. The sleek design allows multiple samples to be dialyzed in the same reservoir.
No additional kits, buoys or accessories are required. The Float-A-Lyzer G2 is packaged dry and ready-to-go with all components included. Simply rinse with water, load the sample with a pipette, and dialyze.
Spectrum Laboratories, Inc. introduces the Spectra/Por® Float-A-Lyzer® G2, the next generation of Ready-to-Use laboratory dialysis devices featuring a Ultra-pure Biotech Cellulose Ester (CE) Membrane
Legal Information
Float-A-Lyzer is a registered trademark of Spectrum Laboratories, Inc.
Spectra/Por is a registered trademark of Spectrum Laboratories, Inc.
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
Z726273Z727253Z727040
description
black, 1 mL, MWCO 3.5-5 kDa
description
black, 10 mL, MWCO 3.5-5 kDa
description
blue, 10 mL, MWCO 100 kDa
description
blue, 1 mL, MWCO 100 kDa
packaging
pack of 12 ea (dry with glycerol)
packaging
pack of 12 ea (dry with glycerol)
packaging
pack of 12 ea (dry with glycerol)
packaging
pack of 12 ea (dry with glycerol)
manufacturer/tradename
Spectrum Labs G235029
manufacturer/tradename
Spectrum Labs G235065
manufacturer/tradename
Spectrum Labs G235071
manufacturer/tradename
Spectrum Labs G235035
material
black
material
black
material
blue
material
blue
pore size
3.5-5 kDa MWCO
pore size
3.5-5 kDa MWCO
pore size
100 kDa MWCO
pore size
100 kDa MWCO
capacity
1 mL
capacity
10 mL
capacity
10 mL
capacity
1 mL
technique(s)
dialysis: suitable
technique(s)
dialysis: suitable
technique(s)
dialysis: suitable
technique(s)
dialysis: suitable
Still not finding the right product?
Explore all of our products under Spectra-Por® Float-A-Lyzer® G2
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)