Przejdź do
Z722804
Hettich® ROTOFIX 32A/Universal 320/320R centrifuge accessories
Rotor, 8 place without tube holders, for ROTOFIX 32A and UNIVERSAL 320
Informacje o tej pozycji
NACRES:
NB.22
UNSPSC Code:
41103900
Manufacturer/tradename:
Hettich 1418
Compatibility:
for use with centrifuge models Rotafix 32A and Universal 320/320R
Parameter:
3,305 max. RCF
Feature:
to hold 8 x 1467, 1468 buckets
feature
to hold 8 x 1467, 1468 buckets
packaging
pack of 1 ea
manufacturer/tradename
Hettich 1418
parameter
3,305 max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models Rotafix 32A and Universal 320/320R
Legal Information
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)