Informacje o tej pozycji
NACRES:
NB.82
UNSPSC Code:
41103900
Manufacturer/tradename:
Hettich 1324
Compatibility:
for use with centrifuge models Rotafix 32A and Universal 320/320R
Parameter:
3,328 max. RCF
Feature:
to hold 4 x 1389, 1490 buckets
feature
to hold 4 x 1389, 1490 buckets
packaging
pack of 1 ea
manufacturer/tradename
Hettich 1324
parameter
3,328 max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models Rotafix 32A and Universal 320/320R
General description
Carriers not included.
Legal Information
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG
Still not finding the right product?
Explore all of our products under Hettich® ROTOFIX 32A/Universal 320/320R centrifuge accessories
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?