Przejdź do
Z722758
Hettich® MIKRO 220/220R centrifuge accessories
for MIKRO 220R Centrifuge, Fixed Angle Rotor, 12 place
Informacje o tej pozycji
NACRES:
NB.22
UNSPSC Code:
41103900
eCl@ss:
32150313
Manufacturer/tradename:
Hettich 1015
Compatibility:
for use with centrifuge models MIKRO 220/220R
Parameter:
4,146 max. RCF
Feature:
to hold 12 x 15 mL tubes
feature
to hold 12 x 15 mL tubes
packaging
pack of 1 ea
manufacturer/tradename
Hettich 1015
parameter
4,146 max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models MIKRO 220/220R
General description
  • compatible with the MIKRO 220R Centrifuge only
Legal Information
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)