Przejdź do
Z721905
Nunc® EasYFlasks™, Nunclon® Δ Surface
Nunclon® Δ surface treatment, 225 cm2 angled neck, vent/close cap, 30/cs
Synonim(y):
cell culture flask, tissue culture flask, tissue culture treated flask, Nunc 159933, surface treated flask
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
30 ea
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
1520,00 zł
Informacje o tej pozycji
NACRES:
NB.22
UNSPSC Code:
41122104
eCl@ss:
32040201
Surface area:
225 cm2
Binding type:
Nunclon Delta surface
Material:
canted neck transparent polystyrene flask, screw top blue high-density polyethylene cap (vent)
Sterility:
sterile; γ-irradiated
Packaging:
case of 30 ea (6 packs x 5 flasks)
Feature:
with cap
1520,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
​
Pomoc techniczna
Pozwól nam pomóc
material
canted neck transparent polystyrene flask, screw top blue high-density polyethylene cap (vent)
sterility
sterile; γ-irradiated
feature
with cap
packaging
case of 30 ea (6 packs x 5 flasks)
manufacturer/tradename
Nunc 159933
technique(s)
cell culture: suitable
flask working volume
70 mL
surface area
225 cm2
fitting
33 mm cap
binding type
Nunclon Delta surface
General description
Enhance cell attachment, growth, and differentiation with these Nunc® EasYFlask™ cell culture-treated flasks. These polystyrene flasks receive the proprietary Nunclon® Delta surface treatment to maximize adhesion for a broad range of cell types. The ergonomic design with angled neck and wide opening allows easy access to the growth surface by pipettes or cell scrapers.
Features and Benefits
  • Nunclon™ Δ certified: a proprietary cell culture surface treatment that offers maximum adhesion for a broad range of cell types.
  • The ergonomic design with angled neck and wide opening allows easy access to the growth surface by pipettes or cell scrapers.
  • Ergonomic design with a 1/3 turn to open, close or vent
  • Low profile to maximize incubator space
  • Volume graduations printed and molded on both sides
  • Slightly angled side walls for complete viewing under a microscope
  • Sterilized by gamma irradiation and certified to be nonpyrogenic
  • Nunclon Delta Certification, including testing on four different cell types, provided in each case
  • Vent/close caps feature a “Y” mark that allows visual verification of vent position.
  • Vent position easily distinguished even when flasks are stacked in incubators.
  • Filter membrane provides effective barrier against micro-organisms.
  • Well-defined, hydrophobic filter membrane offers continuous venting with consistent gas exchange and no lot-to-lot variation.
  • Chemically robust membrane meets USP class VI standards and is produced without animal-derived substances.
Legal Information
EasYFlasks is a trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
F7677F7802TMO159934
description
Nunclon® Δ surface treatment, 225 cm2 angled neck, vent/close cap, 30/cs
description
Nunclon® Δ surface treatment, 175 cm2 angled neck, filter cap, 30/cs
description
Nunclon® Δ surface treatment, 175 cm2 angled neck, vent/close cap, 30/cs
description
polystyrene, capacity 70 mL, Nunclon® Delta surface, sterile, case of 30 ea
manufacturer/tradename
Nunc 159933
manufacturer/tradename
Nunc 159910
manufacturer/tradename
Nunc 159920
manufacturer/tradename
Nunc 159934
binding type
Nunclon Delta surface
binding type
Nunclon Delta surface
binding type
Nunclon Delta surface
binding type
-
sterility
sterile; γ-irradiated
sterility
sterile; γ-irradiated
sterility
sterile; γ-irradiated
sterility
sterile
material
screw top blue high-density polyethylene cap (vent), canted neck transparent polystyrene flask
material
screw top blue high-density polyethylene cap (filter), canted neck transparent polystyrene flask
material
canted neck polystyrene flask, screw top blue high-density polyethylene cap (vent)
material
polystyrene
feature
with cap
feature
with cap
feature
with cap
feature
-
surface area
225 cm2
surface area
175 cm2
surface area
175 cm2
surface area
-
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)