Przejdź do
Z721646
Zone-Free™ Sealing Films
non-sterile
Synonim(y):
qPCR plate seal
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
50 ea
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
784,00 zł
Informacje o tej pozycji
NACRES:
NB.22
UNSPSC Code:
41122410
eCl@ss:
32120613
Manufacturer/tradename:
Excel Scientific ZAF-PE-50
Material:
acrylic adhesive, polyethylene film (top layer), white polypropylene film (sublayer)
Sterility:
non-sterile
Packaging:
pack of 50 ea
784,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
​
Pomoc techniczna
Pozwól nam pomóc
material
acrylic adhesive, polyethylene film (top layer), white polypropylene film (sublayer)
sterility
non-sterile
feature
piercable
packaging
pack of 50 ea
manufacturer/tradename
Excel Scientific ZAF-PE-50
L × W
79.4 mm × 142.9 mm
General description
With a 70 μm polyethylene top layer and inert white polypropylene and acrylic adhesive sublayer, this easy-piercing sealing film allows direct sample recovery with single or multichannel pipettors and robotic probes. The adhesive-free zone above each well prevents adhesive fouling of the pipet tip or probe, ultimately minimizing the amount of adhesive that comes into contact with the sample. Dimensions are 79.4 by 142.9 mm for 96-well plates.
Features and Benefits
  • clear zone above each well is free of adhesive and easily pierceable
  • clear zones and end tabs facilitate well alignment and accurate positioning
  • inert, chemical resistant
  • functional temperature range -40 C to +90 C
Legal Information
Zone-Free is a trademark of Excel Scientific, Inc.
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
Z745195Z734438Z742256
description
non-sterile
description
size 384 wells
description
for qPCR, storage & crystallization, non-sterile
description
for qPCR, storage & crystallization, sterile
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
sterility
sterile
manufacturer/tradename
Excel Scientific ZAF-PE-50
manufacturer/tradename
Excel Scientific ZAF-384-50
manufacturer/tradename
Excel Scientific TSS-RTQ-100
manufacturer/tradename
Excel Scientific TSS-RTQS-50
material
polyethylene film (top layer), acrylic adhesive, white polypropylene film (sublayer)
material
polyethylene , white
material
silicone/adhesive , polyolefin film
material
clear silicone/adhesive , polyolefin film
packaging
pack of 50 ea
packaging
pkg of 1 pack of 50 films
packaging
pack of 100 ea
packaging
pkg of 50 ea
L × W
79.4 mm × 142.9 mm
L × W
-
L × W
76.2 mm × 133.4 mm
L × W
76.2 mm × 133.4 mm
feature
piercable
feature
-
feature
-
feature
-
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)