Merck
Z719617
Sorenson™ low binding standard tips
maximum volume 200 μL, pkg of 96 QuickRack, non-sterile
Synonim(y):
dispensing tip, liquid transfer tip
Informacje o tej pozycji
NACRES:
NB.01
UNSPSC Code:
41121600
Volume:
200 μL
Sterility:
non-sterile
Feature:
barrier: no, graduations
Packaging:
case of 4800 ea (50 racks of 96 tips), pkg of 96 QuickRack, pack of 960 ea (10 racks of 96 tips)
Manufacturer/tradename:
Sorenson 30450T
Pomoc techniczna
Pozwól nam pomóc
material
colorless , polypropylene
sterility
non-sterile
feature
barrier: no, graduations
packaging
case of 4800 ea (50 racks of 96 tips), pkg of 96 QuickRack, pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename
Sorenson 30450T
maximum volume
200 μL
volume
200 μL
General description
Tips feature a new surface technology designed for pipetting substances which tend to stick to the walls of pipette tips or for pipetting expensive reagents. Universal fit for research-grade pipettors.
Legal Information
Sorenson is a trademark of Sorenson BioScience, Inc.
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)