Merck
Z719552
Sorenson™ low binding E tips
maximum volume 10 μL, non-sterile
Synonim(y):
dispensing tip, liquid transfer tip
Informacje o tej pozycji
NACRES:
NB.01
UNSPSC Code:
41121600
Volume:
10 μL
Sterility:
non-sterile
Feature:
barrier: no
Packaging:
case of 4800 ea (50 racks of 96 tips), pack of 96 ea (Racks)
Manufacturer/tradename:
Sorenson 22220T
Pomoc techniczna
Pozwól nam pomóc
material
colorless , polypropylene
sterility
non-sterile
feature
barrier: no
packaging
case of 4800 ea (50 racks of 96 tips), pack of 96 ea (Racks)
manufacturer/tradename
Sorenson 22220T
maximum volume
10 μL
volume
10 μL
General description
Tips feature a new surface technology designed for pipetting substances which tend to stick to the walls of pipette tips or for pipetting expensive reagents. Extended length. Fits Eppendorf ultra-micro pipettors.
Legal Information
Sorenson is a trademark of Sorenson BioScience, Inc.
Still not finding the right product?
Explore all of our products under Sorenson™ low binding E tips
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)