Przejdź do
Z710881
Dri-gauze™ filter liner for Tunair™ shake flask
For 2.5 L flask
Synonim(y):
Dri-gauze Filter, Filter Liner for Shake Flask, Tunair Shake Flask Filter
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
Informacje o tej pozycji
NACRES:
NB.22
UNSPSC Code:
41122104
Material:
polypropylene
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomóc
material
polypropylene
sterility
non-sterile
packaging
case of 5
manufacturer/tradename
IBI SS-3019
General description
The Dri-Gauze™ filter is 0.22 μm and constructed of nitrocellulose. Each filter can be used 10 – 12 times. The Dri-Gauze filters can be autoclaved in the cap. It consists of five dri-gauze filter cap linings to accommodate a 300 ml flask. The filter lining in the cap assembly can be replaced by pinching the flanges of the inner-closure shell until they snap loose. Then pull them apart and remove the used lining. The lining is replaced by sandwiching it between two parts of the cap and placing the cap back together. Ensure the flanges from the inner piece snap into the mated grooves in the outer piece, while reassembling. This makes the cap assembly stays intact during use.
Legal Information
DRI-GAUZE is a trademark of IBI Scientific
Tunair is a trademark of IBI Scientific
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
Z710873Z710814Z710857
description
For 2.5 L flask
description
For 300 mL flask
description
flask volume 2,500 mL, polypropylene, half baffle
description
For 300 mL flask
material
polypropylene
material
polypropylene
material
wide-mouth , polypropylene , straight neck
material
silicon
packaging
case of 5
packaging
case of 5
packaging
case of 4
packaging
case of 5
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
manufacturer/tradename
IBI SS-3019
manufacturer/tradename
IBI SS-3018
manufacturer/tradename
IBI SS-8002
manufacturer/tradename
IBI SS-3016
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)