Przejdź do
Z710024
Sartorius pipette tips
volume range 50-1200 μL, standard, refill, non-sterile
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
Informacje o tej pozycji
NACRES:
NB.01
UNSPSC Code:
41121600
Volume:
50—1250 μL
Sterility:
non-sterile
Feature:
DNase free, RNase free, barrier: no, endotoxin free
Packaging:
pack of 96 ea (Refil rack)
Manufacturer/tradename:
Sartorius 791202
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomóc
material
colorless
sterility
non-sterile
feature
DNase free, RNase free, barrier: no, endotoxin free
packaging
pack of 96 ea (Refil rack)
manufacturer/tradename
Sartorius 791202
maximum volume
1.2 mL
volume range
50-1200 μL
volume
50—1250 μL
General description
The non-filtered Optifit Tips are designed for best possible fitting and sealing to secure the highest possible accuracy and precision. These pipette tips enable ergonomic and light tip attachment.
Optifit Tips are available in various packaging types for all your needs:
  • Single Tray racks – purity certified tip racks
  • Refill Packs - purity certified and ecological refill packages
  • Refill Towers - space-saving and ecological refill packages
  • Flexibulk boxes - space-saving, ecological and economical bulk boxes
  • Standard bulk – economical bulk boxes
While Optifit Tip trays are colour coded for easy matching with a corresponding, colour coded Sartorius pipette, they do also match with most other pipette due to their universal design.
Strict quality standards, ISO 9001 and ISO 13485, are followed from tip R&D to production and delivery. The purity certified tips are manufactured in ISO 8 classified clean room conditions to avoid any contaminations and are tested for DNase, RNase and endotoxins per each tip lot. Pre-sterilised tips are also available for the purest laboratory work. Sartorius offers a selection of pipette tips, extended and large volume tips, for special needs.
Other Notes
Browse all pipette tips
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
Z710016Z757756Z757713
description
volume range 50-1200 μL, standard, refill, non-sterile
description
volume range 50-1200 μL, standard, racked, sterile
description
volume range 50-1200 μL, standard, refill, sterile
description
volume range 0.5-300 μL, standard, refill, sterile
volume
50—1250 μL
volume
50—1250 μL
volume
50—1250 μL
volume
0.5—300 μL
sterility
non-sterile
sterility
sterile
sterility
sterile
sterility
sterile
manufacturer/tradename
Sartorius 791202
manufacturer/tradename
Sartorius 791201
manufacturer/tradename
Sartorius 791203
manufacturer/tradename
Sartorius 790203
feature
endotoxin free, RNase free, barrier: no, DNase free
feature
DNase free, barrier: no, RNase free, endotoxin free
feature
barrier: no, endotoxin free, RNase free, DNase free
feature
RNase free, barrier: no, DNase free, endotoxin free
material
colorless
material
colorless
material
colorless
material
colorless
packaging
pack of 96 ea (Refil rack)
packaging
pack of 96 ea (Racked)
packaging
pack of 96 ea (Racked)
packaging
pack of 96 ea (Racked)
Still not finding the right product?
Explore all of our products under Sartorius pipette tips
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)