Merck
Z708267
Slide mailer, 2 place
assorted colors, natural, red, yellow
Informacje o tej pozycji
NACRES:
MA.01
UNSPSC Code:
41123302
Manufacturer/tradename:
Heathrow Scientific HS15983A
Feature:
Holds 2 x slides 76x26 mm
Material:
polypropylene
Pomoc techniczna
Pozwól nam pomóc
material
polypropylene
feature
Holds 2 x slides 76x26 mm
manufacturer/tradename
Heathrow Scientific HS15983A
size
93x68x50 mm
color
assorted colors, natural, red, yellow
General description
These 2-place slide protectors hold standard slides (3 × 1 in. or 25 × 75 mm) securely. With large, easy to secure snaps our mailers provide safe storage for valuable slides. The bright neon colors bring some fun to the lab and come in handy for color-coding
Still not finding the right product?
Explore all of our products under Slide mailer, 2 place
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)