Przejdź do
Z694479
Nanofiber chamber slides
8 chamber slide, random oriented nanofibers
Synonim(y):
3D, 3D Cell Culture, 3d plates, tissue culture plates, Nanofiber Solutions 0801
Wybierz wielkość
Zmień widok
Do Państwa/SKUDostępnośćCena netto
1 ea
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
1020,00 zł
867,00 zł
Informacje o tej pozycji
NACRES:
NB.22
UNSPSC Code:
41121800
Sterility:
non-sterile
867,00 zł
Cena katalogowa1020,00 złZaoszczędź 15%
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
Pomoc techniczna
Pozwól nam pomóc
sterility
non-sterile
manufacturer/tradename
Nanofiber Solutions 801
suitability
suitable for (Tissue Engineering, Cancer Research)
General description
Nanofiber cell culture dishes, multiwell plates, chamber slides and well plate inserts combine the best of 3D and 2D cell culture. The nanofibers are optically transparent to allow for live-cell imaging and real time quantification of cell mobility. The nanofibers mimic the 3D topography in vivo, and can be easily coated with ECM proteins for Cancer Research. The Cells can be simply removed for protein or gene analysis.
The nanofibers have higher expansion rates, better differentiation control, and has a synthetic surface with no animal contamination, ideal for stem cells research and immunohistochemistry staining. Nanofiber scaffolds can be easily shaped or sized, made from nearly any synthetic or natural polymer for customized tissue engineering. They can then be implanted in vivo or used in vitro. The standard plate dimensions available will work with current automated equipment for high throughput drug discovery needs.
The cell culture dishes, plates, chamber slides and inserts are available with either randomly oriented fibers or aligned fibers.
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
Z694576Z694614Z694584
description
8 chamber slide, random oriented nanofibers
description
8 chamber slide, aligned nanofibers
description
for 12 well plates
description
diam. 60 mm
manufacturer/tradename
Nanofiber Solutions 801
manufacturer/tradename
Nanofiber Solutions 802
manufacturer/tradename
Nanofiber Solutions 121202
manufacturer/tradename
Nanofiber Solutions 6002
sterility
non-sterile
sterility
non-sterile
sterility
sterile; electron beam irradiated
sterility
sterile
suitability
suitable for (Tissue Engineering, Cancer Research)
suitability
suitable for (Tissue Engineering, Cancer Research)
suitability
suitable for (Tissue Engineering, Cancer Research)
suitability
suitable for (Tissue Engineering, Cancer Research)
Still not finding the right product?
Explore all of our products under Nanofiber chamber slides
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)