Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 429,00 zł |
Informacje o tej pozycji
429,00 zł
material
(polycaprolactone (PCL)), flat clear wells, transparent polycaprolactone (PCL)
sterility
sterile
feature
lid
manufacturer/tradename
Nanofiber Solutions 9601
technique(s)
cell culture: suitable
size
96 wells
suitability
suitable for (Tissue Engineering, Cancer Research)
application(s)
3D cell culture
binding type
non-treated surface
General description
The nanofibers have higher expansion rates, better differentiation control, and has a synthetic surface with no animal contamination, ideal for stem cells research and immunohistochemistry staining. Nanofiber scaffolds can be easily shaped or sized, made from nearly any synthetic or natural polymer for customized tissue engineering. They can then be implanted in vivo or used in vitro. The standard plate dimensions available will work with current automated equipment for high throughput drug discovery needs.
The cell culture dishes, plates and inserts are available with either randomly oriented fibers or aligned fibers.
For fluorescent imaging applications, we recommend using the Nanofiber Solutions chamber slides.
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description size 96 wells | description size 384 wells | description size 6 wells | description size 24 wells |
| sterility sterile | sterility sterile | sterility sterile | sterility sterile |
| manufacturer/tradename Nanofiber Solutions 9601 | manufacturer/tradename Nanofiber Solutions 38401 | manufacturer/tradename Nanofiber Solutions 601 | manufacturer/tradename Nanofiber Solutions 2401 |
| feature lid | feature lid | feature lid | feature lid |
| binding type non-treated surface | binding type non-treated surface | binding type non-treated surface | binding type non-treated surface |
| material (polycaprolactone (PCL)), transparent polycaprolactone (PCL) , flat clear wells | material transparent polycaprolactone (PCL) , flat bottom clear wells, (polycaprolactone (PCL)) | material (polycaprolactone (PCL)), flat bottom clear wells, transparent polycaprolactone (PCL) | material flat bottom clear wells, (polycaprolactone (PCL)), transparent polycaprolactone (PCL) |
| suitability suitable for (Tissue Engineering, Cancer Research) | suitability suitable for (Tissue Engineering, Cancer Research) | suitability suitable for (Tissue Engineering, Cancer Research) | suitability suitable for (Tissue engineering, cancer research) |
Still not finding the right product?
Explore all of our products under Nanofiber multiwell plate, with random oriented nanofibers
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?