Wybierz wielkość
| Do Państwa/SKU | Dostępność | Cena netto |
|---|---|---|
450 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 3580,00 zł |
Informacje o tej pozycji
3580,00 zł
material
colorless tube, polyethylene cap, polystyrene tube, round bottom, screw top cap
sterility
sterile
feature
neck thread external
packaging
pack of 450 ea
manufacturer/tradename
Nunc 156758
L × O.D.
110 mm × 16 mm
capacity
3 mL
General description
Features and Benefits
- Polystyrene tubes, polyethylene caps
- 5.5cm2 culture area for grow of adherent cells
- Allows easy microscopy; a coverslip can be used in the tube
- Medium retained in the flat-sided tube in horizontal position
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description sterile | description Nunclon® Δ surface treatment, 175 cm2 angled neck, vent/close cap, 30/cs | description Nunclon® Δ surface treatment, 25 cm2 angled neck, filter cap, 200/cs | description capacity 1.8 mL, External thread, round bottom (self-standing), sterile; irradiated |
| manufacturer/tradename Nunc 156758 | manufacturer/tradename Nunc 159920 | manufacturer/tradename Nunc 156367 | manufacturer/tradename Nunc 375418 |
| sterility sterile | sterility sterile; γ-irradiated | sterility sterile; γ-irradiated | sterility sterile; irradiated |
| material colorless tube, polystyrene tube, screw top cap, polyethylene cap, round bottom | material canted neck polystyrene flask, screw top blue high-density polyethylene cap (vent) | material canted neck transparent polystyrene flask, screw top blue high-density polyethylene cap (filter) | material colorless polyethylene screw cap, colorless tube, polypropylene tube, round bottom (self-standing) |
| feature neck thread external | feature with cap | feature with cap | feature External thread, with writing surface |
| packaging pack of 450 ea | packaging case of 30 ea (6 packs x 5 flasks) | packaging case of 200 ea (20 packs x 10 flasks) | packaging pack of 450 ea |
| capacity 3 mL | capacity - | capacity - | capacity 1.8 mL |
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?