Przejdź do
Z688320
LightSafe centrifuge tubes
capacity 15 mL, conical polypropylene, sterile
Informacje o tej pozycji
NACRES:
NB.22
UNSPSC Code:
41121703
eCl@ss:
32040302
Capacity:
15 mL
Material:
black tube, cap (attached), conical polypropylene
Sterility:
sterile
Packaging:
pack of 50
Feature:
RCF 8,400 × g
material
black tube, cap (attached), conical polypropylene
sterility
sterile
feature
RCF 8,400 × g
packaging
pack of 50
manufacturer/tradename
Cole Parmer 634430
capacity
15 mL
General description
Disposable polypropylene tubes and caps with recessed lid to prevent leaking. Frosted writing surface allows for permanent identification and labeling. Autoclavable to 122°C and can be stored in freezers to -100°C. RCF values: 15 mL, 8,400 g/50 mL, 10,000 g
Still not finding the right product?
Explore all of our products under LightSafe centrifuge tubes
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)