Merck
Z688169
HandEvac handheld aspirator
complete operator set
Synonim(y):
Handheld Aspirator, Handheld Vacuum Aspirator, Portable Aspirator
Informacje o tej pozycji
NACRES:
NB.22
UNSPSC Code:
41105100
Pomoc techniczna
Pozwól nam pomóc
manufacturer/tradename
Argos 1305032
technique(s)
cell culture: suitable
General description
Rapidly remove supernatant from a variety of cell culture or multi-well plates and dishes!
This ergonomic, hand-held aspirator can be used with pasteur pipettes, plastic aspiration pipettes or a variety of adapters for specific applications. A pressure-sensitive button on the top of the unit controls the specific amount of vacuum applied. This prevents the accidental aspiration of cells or supernatant associated with normal vacuum techniques.
Use with standard in-house vacuum source or with the M-Vac or E-Vac aspiration systems. Autoclavable unit is supplied with a single-channel adapter for disposable pipette tips.
Includes: EV526, EV501, EV520 and EV503
Still not finding the right product?
Explore all of our products under HandEvac handheld aspirator
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)