Przejdź do zawartości
Z659738
Bandelin SONOREX Ultrasonic bath accessories
insert tub with lid for Z659673 and Z659029
Synonim(y):
Bandelin 240, holder, retainer, sonification holder, sonification tub sonifier insert
Informacje o tej pozycji
NACRES:
NB.74
UNSPSC Code:
41103706
Pomoc techniczna
Pozwól nam pomóc
material
polyethylene
description
Bandelin KW 5
packaging
pack of 1 ea
manufacturer/tradename
Bandelin 240
availability
not available in USA and Canada
L × W × D
254 mm × 96 mm × 130 mm
General description
For use with chemically aggressive liquids, removing chemically aggressive or abrasive contamination or sonicating sample liquids, insert cups are used (indirect sonication). Because nothing may be placed on the bottom of the ultrasonic bath.
Features and Benefits
  • for chemically aggressive liquids
  • protects the stainless steel tub
Legal Information
SONOREX is a trademark of Bandelin Electronic GmbH & Co. KG
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)