Merck
Z659487
Bandelin SONOREX™ Ultrasonic bath accessories
tank lid to fit Z659460
Synonim(y):
Bandelin 218, lid, protective lid, tank cover, ultrasonic cover
Informacje o tej pozycji
NACRES:
NB.74
UNSPSC Code:
41103706
Pomoc techniczna
Pozwól nam pomóc
material
stainless steel
description
Bandelin D 08
packaging
pack of 1 ea
manufacturer/tradename
Bandelin 218
General description
Stainless steel lid for an ultrasonic bath protects the bath liquid from external contamination and prevents unpleasant odors from the cleaning or disinfection agents added. In addition, the noise emitted when the ultrasonic cleaner is in operation, which may be perceived as annoying, is reduced. The lids are designed in such a way that any condensation that occurs is directed back into the bath.
Features and Benefits
  • Protection of the bath fluid from contamination
  • Sound-absorbing
  • Shortening the heating phase
Legal Information
SONOREX is a trademark of Bandelin Electronic GmbH & Co. KG
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)