Przejdź do
Z657867
Hettich® centrifuge adapters
for 4.5-5 mL blood/urine tubes (11x92 mm) and 7ml round bottom tubes (12x100 mm)
Informacje o tej pozycji
NACRES:
NB.22
UNSPSC Code:
41103911
Compatibility:
for use with 1427 (bucket)
Manufacturer/tradename:
Hettich 5230
description
for UNIVERSAL 320/320R and ROTINA 38/38R
feature
Holds 7 mL tubes
packaging
pack of 1 ea
manufacturer/tradename
Hettich 5230
technique(s)
centrifugation: suitable
compatibility
for use with 1427 (bucket)
Legal Information
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
Z657883Z657875Z653403
manufacturer/tradename
Hettich 5230
manufacturer/tradename
Hettich 5233
manufacturer/tradename
Hettich 5231
manufacturer/tradename
Hettich 1451
compatibility
for use with 1427 (bucket)
compatibility
for use with 1427 (bucket)
compatibility
for use with 1427 (bucket)
compatibility
for use with 5615, 4246, 1620A, 1720, 1792, 4794 (rotors)
description
for UNIVERSAL 320/320R and ROTINA 38/38R
description
-
description
for ROTINA 38/38R and UNIVERSAL 320/320R
description
for ROTINA 38/38R, ROTOFIX 32A and UNIVERSAL 320/320R
technique(s)
centrifugation: suitable
technique(s)
centrifugation: suitable
technique(s)
centrifugation: suitable
technique(s)
centrifugation: suitable
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
feature
Holds 7 mL tubes
feature
Holds 50 mL tubes
feature
Holds 15 mL tubes
feature
Holds 15 mL tubes
Still not finding the right product?
Explore all of our products under Hettich® centrifuge adapters
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)