Przejdź do
Z653365
Hettich® centrifuge buckets
for ROTOFIX 32A and UNIVERSAL 320/320R, bucket/carrier for rotors 1624; 1726
Informacje o tej pozycji
NACRES:
NB.22
UNSPSC Code:
41103910
Compatibility:
for use with rotors 1624 (Z601527) and 1726 (Z631108)
Manufacturer/tradename:
Hettich 1741
Feature:
Holds 10 x 4.9 mLblood collection/urine vessels
feature
Holds 10 x 4.9 mLblood collection/urine vessels
manufacturer/tradename
Hettich 1741
availability
available only in EU, not available in USA and Canada
technique(s)
centrifugation: suitable
compatibility
for use with rotors 1624 (Z601527) and 1726 (Z631108)
General description
  • compatible with rotors 1624 (Z601527) and 1726 (Z631108)
Legal Information
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)