Merck
Z637459
BRAND® microtube cap inserts, PP
white
Synonim(y):
cap insert for microtube, cap insert for screw caps tube
Informacje o tej pozycji
NACRES:
NB.11
UNSPSC Code:
41121702
eCl@ss:
32040301
Color:
white
Pomoc techniczna
Pozwól nam pomóc
manufacturer/tradename
BRAND 780720
color
white
General description
The BRAND® Microtube Cap Inserts are designed for use with microtubes featuring attached or bulk screw caps that are non-tamper evident. Made from durable polypropylene (PP), these inserts provide a secure closure for sample integrity. They are suitable for a wide range of applications, with an operating temperature range from -196 °C to +121 °C, making them ideal for both cryogenic and high-temperature environments. These cap inserts are essential for ensuring reliable storage and handling of samples in various laboratory settings.
Other Notes
BR780720 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Still not finding the right product?
Explore all of our products under BRAND® microtube cap inserts, PP
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)