Przejdź do
Z617849
Greiner PS tube
capacity × diam. × L 14 mL × 18 mm × 95 mm
Synonim(y):
Greiner 191161, culture tube, plastic tube, round bottom culture tube, tube
Informacje o tej pozycji
NACRES:
NB.22
UNSPSC Code:
41121706
material
cap (Two-Position Vent Stopper), clear tube, polystyrene , polystyrene tube, round bottom tube, round bottom, straight neck
sterility
sterile
feature
RNase and DNase free, neck thread push fit
packaging
pack of 750 ea
manufacturer/tradename
Greiner 191161
technique(s)
cell culture: suitable
capacity × diam. × L
14 mL × 18 mm × 95 mm
General description
  • Wysoka przezroczystość
  • Okrągłe dno z dwupozycyjnym korkiem odpowietrzającym
Greiner Bio-One culture tubes with two-stage vent cap The polystyrene material is ideal for optical measurements due to its high clarity. and is free of detectable DNase, RNase or human DNA, pyrogen-free.
Legal Information
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)