Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
25 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 976,00 zł |
Informacje o tej pozycji
976,00 zł
material
polycarbonate housing, polyethersulfone membrane
sterility
non-sterile
feature
holdup volume 10 μL
packaging
package of 25 ea
manufacturer/tradename
Sartorius VS0231
parameter
8000 × g max. centrifugal force
dead stop volume
8 μL
membrane area
1.2 cm2
tube size
~15 mL
working volume
2 mL
pore size
50 kDa MWCO
General description
- Vivaspin centrifugal concentrators offer the optimal solution to any concentration or buffer exchange application with their broad range of volume capacities and MWCOs.
- Highest flow rates and recoveries are achieved due to their particular vertical membrane design.
- Incomparably safe sample handling and convenience is assured by the patented dead-stop technology allowing direct recovery of the concentrate without further re-spin.
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description MWCO 50 kDa | description MWCO 30 kDa | description MWCO 100 kDa | description MWCO 5 kDa |
| parameter 8000 × g max. centrifugal force | parameter 8000 × g max. centrifugal force | parameter 8000 × g max. centrifugal force | parameter 8000 × g max. centrifugal force |
| pore size 50 kDa MWCO | pore size 30 kDa MWCO | pore size 100 kDa MWCO | pore size 5 kDa MWCO |
| sterility non-sterile | sterility non-sterile | sterility non-sterile | sterility non-sterile |
| manufacturer/tradename Sartorius VS0231 | manufacturer/tradename Sartorius VS0221 | manufacturer/tradename Sartorius VS0241 | manufacturer/tradename Sartorius VS0211 |
| material polycarbonate housing, polyethersulfone membrane | material polycarbonate housing, polyethersulfone membrane | material polycarbonate housing, polyethersulfone membrane | material polyethersulfone membrane, polycarbonate housing |
| working volume 2 mL | working volume 2 mL | working volume 2 mL | working volume 2 mL |
Still not finding the right product?
Explore all of our products under Vivaspin 2 centrifugal concentrators
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?