Przejdź do
Z601535
Hettich® centrifuge buckets
for ROTOFIX 32A, ROTINA 38/38R and UNIVERSAL 320/320R Centrifuges, bucket/carrier for rotors 1624; 1726
Informacje o tej pozycji
NACRES:
NB.22
UNSPSC Code:
41103910
Compatibility:
for use with rotors 1624 (Z601527) and 1726 (Z631108)
Manufacturer/tradename:
Hettich 1742
Feature:
Holds 7 x 15 mLtubes
feature
Holds 7 x 15 mLtubes
packaging
pack of 4 ea
manufacturer/tradename
Hettich 1742
availability
available only in EU, not available in USA and Canada
technique(s)
centrifugation: suitable
compatibility
for use with rotors 1624 (Z601527) and 1726 (Z631108)
General description
  • compatible with rotors 1624 (Z601527) and 1726 (Z631108)
Legal Information
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)