Merck
Z601470
Hettich® centrifuge buckets
for ROTOFIX 32A, ROTINA 38/38R and UNIVERSAL 320/320R, bucket/carrier for rotor 1624; 1726
Informacje o tej pozycji
NACRES:
NB.22
UNSPSC Code:
41103910
eCl@ss:
32110390
Compatibility:
for use with rotors 1624 (Z601527) and 1726 (Z631108)
Manufacturer/tradename:
Hettich 1372
Feature:
Holds 17 x 5mltubes
Pomoc techniczna
Pozwól nam pomóc
feature
Holds 17 x 5mltubes
packaging
pack of 4 ea
manufacturer/tradename
Hettich 1372
availability
available only in EU, not available in USA and Canada
technique(s)
centrifugation: suitable
compatibility
for use with rotors 1624 (Z601527) and 1726 (Z631108)
General description
  • compatible with rotors 1624 (Z601527) and 1726 (Z631108)
Legal Information
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)