Informacje o tej pozycji
NACRES:
NB.82
UNSPSC Code:
41103900
Manufacturer/tradename:
Hettich 1016
Compatibility:
for use with centrifuge models MIKRO 220/220R
Parameter:
4,025 max. RCF
Feature:
capacity 7 ‑ 50 mL, to hold 6 x 50 mL tubes
feature
capacity 7 ‑ 50 mL, to hold 6 x 50 mL tubes
packaging
pack of 1 ea
manufacturer/tradename
Hettich 1016
parameter
4,025 max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models MIKRO 220/220R
General description
- compatible with the MIKRO 220R Centrifuge only
Legal Information
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG
Still not finding the right product?
Explore all of our products under Hettich® MIKRO 220/220R centrifuge accessories
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?