Informacje o tej pozycji
NACRES:
NB.01
UNSPSC Code:
41121600
Volume:
10 μL
Sterility:
non-sterile
Packaging:
pack of 200 ea (Bag)
Manufacturer/tradename:
Sorenson 1511
Pomoc techniczna
Pozwól nam pomócmaterial
colorless , polypropylene
sterility
non-sterile
packaging
pack of 200 ea (Bag)
manufacturer/tradename
Sorenson
maximum volume
10 μL
volume
10 μL
General description
Low capacity improves accuracy with small sample volumes by reducing dead space.
Application
For loading sequencing gels 0.6 mm or thicker.
Other Notes
Browse all pipette tips
Still not finding the right product?
Explore all of our products under Ultramicro pipette tips
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?