Zmień widok
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
NACRES:
NB.22
UNSPSC Code:
41123102
eCl@ss:
34220503
Material:
blue
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomócmaterial
blue
technique(s)
dialysis: suitable
size
70 mm
General description
Clamping closures gently lock over dialysis tubing, providing a leakproof seal without knots. Can be used with cellulose ester, regenerated cellulose, and PVDF dialysis membranes. Made entirely of Nylon 66, closures have an operating temperature range of 0-90°C, and can be sterilized with ethylene oxide, γ-irradiation, ethanol, or formalin.
Different colors can be used to code samples. For best results, select a closure 4-10 mm wider than the flat width of the dialysis tubing.
Different colors can be used to code samples. For best results, select a closure 4-10 mm wider than the flat width of the dialysis tubing.
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description blue, size 70 mm | description white, size 70 mm | description red, size 70 mm | description blue, size 50 mm |
| material blue | material white | material red | material blue |
| technique(s) dialysis: suitable | technique(s) dialysis: suitable | technique(s) dialysis: suitable | technique(s) dialysis: suitable |
| size 70 mm | size 70 mm | size 70 mm | size 50 mm |
Still not finding the right product?
Explore all of our products under Dialysis tubing closures
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?