Merck
Z371017
Dialysis tubing closures
red, size 110 mm
, aby wyświetlić ceny organizacyjne i kontraktowe.
Wybierz wielkość
Zmień widok
Informacje o tej pozycji
NACRES:
NB.22
UNSPSC Code:
41123102
Material:
red
Pomoc techniczna
Pozwól nam pomóc
material
red
size
110 mm
General description
Clamping closures gently lock over dialysis tubing, providing a leakproof seal without knots. Can be used with cellulose ester, regenerated cellulose, and PVDF dialysis membranes. Made entirely of Nylon 66, closures have an operating temperature range of 0-90°C, and can be sterilized with ethylene oxide, γ-irradiation, ethanol, or formalin.
Different colors can be used to code samples. For best results, select a closure 4-10 mm wider than the flat width of the dialysis tubing.
Still not finding the right product?
Explore all of our products under Dialysis tubing closures
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)