Merck
Z370762
Scienceware® Holdfast™ graduated cylinders with handle
volume 1000 mL
Synonim(y):
284611000, F28461-1000, flask with handles, graduated measuring cylinder, liquid measuring device, measuring cylinder
Informacje o tej pozycji
NACRES:
NB.43
UNSPSC Code:
41121805
Volume measuring range:
1000 mL
Material:
polypropylene
Packaging:
pack of 1 ea
Pomoc techniczna
Pozwól nam pomóc
material
polypropylene
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F28461-1000
volume measuring range
1000 mL
General description
Autoclavable polypropylene with double scale and stable octagon base. The handle provides a safe, solid grip for more controlled pouring from these large-sized cylinders.
Legal Information
Holdfast is a trademark of Bel-Art Products, a division of SP
Scienceware is a registered trademark of Bel-Art Products, a division of SP
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)