Przejdź do
Z367729
Low-profile roller
low-profile roller, AC/DC input 230 V AC, European 2-pin plug
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
Informacje o tej pozycji
NACRES:
NB.73
UNSPSC Code:
41103811
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomóc
feature
CE compliant
manufacturer/tradename
IBI
L × W × H
362 mm × 381 mm × 60 mm
General description
Weather-proof roller operates on benchtop, in dry or humid incubators, in cold rooms to 0°C, and even in ovens to 65°C. Use for cell culture with standard roller bottles and in filter blot applications with hybridization bottles (below). Unit is supplied with six gear-driven rollers, to rotate three large bottles at variable speed, 0.25−8 rpm. Rollers can be moved closer together to accommodate small bottles and tubes, and up to five more rollers can be added; as many as thirty 10 × 75 mm tubes can be turned at 2−67 rpm. Unit is powered by a cool-running 24 VDC motor. Thin, flat AC power cord (US only) reaches external outlet without damaging incubator or refrigerator gasket.
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
Z367737Z742001Z742003
description
low-profile roller, AC/DC input 230 V AC, European 2-pin plug
description
extra roller
description
Lab Start-Up Kit
description
-
manufacturer/tradename
IBI
manufacturer/tradename
IBI ACCRLSR10
manufacturer/tradename
IBI ROLLS0001
manufacturer/tradename
IBI ACCRL0002
L × W × H
362 mm × 381 mm × 60 mm
L × W × H
362 mm × 381 mm × 60 mm
L × W × H
-
L × W × H
-
feature
CE compliant
feature
-
feature
Includes Five Replacement Rollers (Z367737) and bag of assorted o-rings
feature
-
Still not finding the right product?
Explore all of our products under Low-profile roller
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)