Zmień widok
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
NACRES:
NB.12
UNSPSC Code:
41122800
Manufacturer/tradename:
BRAND 388050
Feature:
Holds 2 x Imhoff cones
Material:
PMMA
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomócmaterial
PMMA
feature
Holds 2 x Imhoff cones
manufacturer/tradename
BRAND 388050
L × W × H
300 mm × 130 mm × 315 mm
General description
The BRAND® Imhoff Cone Rack is designed to securely hold two Imhoff sedimentation cones made of plastic, providing a practical solution for sedimentation processes. Constructed from a combination of PMMA and PP, this rack offers durability and stability while remaining lightweight. Its compact design makes it easy to carry, even when the cones are filled, facilitating convenient transport and handling in laboratory settings. This essential accessory enhances the efficiency of sedimentation experiments, ensuring that your cones are securely supported and easily accessible.
Other Notes
BR388050 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description PMMA | description to hold, holds 3 cones | description joint: ST/NS 7/16 | description conical bottom |
| feature Holds 2 x Imhoff cones | feature to hold holds 3 cones | feature autoclavable | feature graduations: no |
| manufacturer/tradename BRAND 388050 | manufacturer/tradename Bel-Art F38993-0003 | manufacturer/tradename BRAND | manufacturer/tradename BRAND 114740 |
| material PMMA | material - | material - | material polystyrene tube, conical bottom, colorless tube |
| L × W × H 300 mm × 130 mm × 315 mm | L × W × H - | L × W × H - | L × W × H - |
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?