Informacje o tej pozycji
NACRES:
NB.43
UNSPSC Code:
41121804
eCl@ss:
32040501
Capacity:
500 mL
Material:
borosilicate glass 3.3 , round bottom flask, single-neck
Joint:
joint: ST/NS29/32
Packaging:
case of 1
Pomoc techniczna
Pozwól nam pomócmaterial
borosilicate glass 3.3 , round bottom flask, single-neck
joint
joint: ST/NS29/32
agency
according to DIN 12 348
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
case of 1
manufacturer/tradename
Lenz 3.0029.58
capacity
500 mL
Legal Information
Lenz is a registered trademark of Lenz GmbH
Still not finding the right product?
Explore all of our products under Lenz® round bottom flasks
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?