Merck
Z329800
BRAND® wash bottle, wide-mouth, labelled, LDPE
capacity 500 mL, label, no label, without venting valve
Informacje o tej pozycji
NACRES:
NB.17
UNSPSC Code:
41103209
eCl@ss:
32040107
Pomoc techniczna
Pozwól nam pomóc
material
low-density polyethylene bottle, natural bottle, round bottle, wide-mouth bottle, yellow closure
feature
label no label, with cap, without venting valve
packaging
pack of 5 ea
manufacturer/tradename
BRAND 144089
capacity
500 mL
General description
Flexible, high-quality wash bottles with integral wash steam and sealing cone in the closure. Large opening for convenient filling. Label with designation of contents and general safety statements (with CAS no. and NFPA codes).
Bottles with venting valve (metal ball) are for distilled water and organic solvents only; to avoid dropping caused by overpressure.
Other Notes
BR144089 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)