Przejdź do zawartości
Z329029
BRAND® magnetic stirring bar, PTFE, cylindrical with pivot ring
L 45 mm, bar diam. 8 mm
Synonim(y):
cylindrical stir bar, magnetic bar, magnetic stir bar, spinbar, stir bar
Informacje o tej pozycji
NACRES:
NB.72
UNSPSC Code:
41103806
Material:
cylindrical PTFE-coated
Manufacturer/tradename:
BRAND 137432
Packaging:
pack of 10
Pomoc techniczna
Pozwól nam pomóc
material
cylindrical PTFE-coated
packaging
pack of 10
manufacturer/tradename
BRAND 137432
L
45 mm
bar diam.
8 mm
ring diam.
10 mm
General description
For vessels with slightly uneven bottoms. The pivot ring reduces the contact surface and enables the stirrer to adopt the optimum stirring position.
Other Notes
BR137432 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)