Przejdź do
Z328790
BRAND® magnetic stirring bar, PTFE, cylindrical
L 30 mm, bar diam. 6 mm
Synonim(y):
PTFE, Stirring rods
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
Informacje o tej pozycji
NACRES:
NB.72
UNSPSC Code:
41103806
eCl@ss:
32100101
Material:
PTFE-coated , cylindrical
Manufacturer/tradename:
BRAND 137125
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomóc
Quality Segment
material
PTFE-coated , cylindrical
manufacturer/tradename
BRAND 137125
L
30 mm
bar diam.
6 mm
General description
BRAND® magnetic cylindrical stirring bars are expertly designed for optimal mixing performance, featuring a smooth, round shape that enhances stirring efficiency. Their Alnico V magnetic cores are protected by a robust PTFE (polytetrafluoroethylene) coating, ensuring durability and resistance to chemical damage. These stirring bars are particularly effective for use in large volumes or thick mixtures, making them ideal for a variety of laboratory applications.
Features and Benefits
  • BRAND® magnetic cylindrical stirring bars are round with a smooth surface.
  • The Alnico V magnetic cores are encapsulated with high-grade, crack-resistant PTFE (polytetrafluoroethylene) coating.
  • Ideal for large volumes or highly viscous mixtures.
Other Notes
BR137125 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
Z328650Z328669Z328766
description
L 30 mm, bar diam. 6 mm
description
L 10 mm, bar diam. 3 mm
description
L 10 mm, bar diam. 6 mm
description
L 20 mm, bar diam. 6 mm
manufacturer/tradename
BRAND 137125
manufacturer/tradename
BRAND 137108
manufacturer/tradename
BRAND 137113
manufacturer/tradename
BRAND 137115
material
cylindrical , PTFE-coated
material
cylindrical , PTFE-coated
material
PTFE-coated , cylindrical
material
PTFE-coated , cylindrical
bar diam.
6 mm
bar diam.
3 mm
bar diam.
6 mm
bar diam.
6 mm
Quality Level
200
Quality Level
200
Quality Level
200
Quality Level
200
L
30 mm
L
10 mm
L
10 mm
L
20 mm
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)