Zmień widok
| Do Państwa/SKU | Dostępność | Cena netto |
|---|---|---|
10 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 175,00 zł |
Informacje o tej pozycji
UNSPSC Code:
41103806
NACRES:
NB.72
eCl@ss:
32100100
Material:
PTFE-coated , cylindrical
Manufacturer/tradename:
BRAND 137109
175,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
Pomoc techniczna
Pozwól nam pomócQuality Segment
material
PTFE-coated , cylindrical
manufacturer/tradename
BRAND 137109
L
13 mm
bar diam.
3 mm
General description
BRAND® magnetic cylindrical stirring bars are expertly designed for optimal mixing performance, featuring a smooth, round shape that enhances stirring efficiency. Their Alnico V magnetic cores are protected by a robust PTFE (polytetrafluoroethylene) coating, ensuring durability and resistance to chemical damage. These stirring bars are particularly effective for use in large volumes or thick mixtures, making them ideal for a variety of laboratory applications.
Features and Benefits
- BRAND® magnetic cylindrical stirring bars are round with a smooth surface.
- The Alnico V magnetic cores are encapsulated with high-grade, crack-resistant PTFE (polytetrafluoroethylene) coating.
- Ideal for large volumes or highly viscous mixtures.
Other Notes
BR137109 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description L 13 mm, bar diam. 3 mm | description L 15 mm, bar diam. 6 mm | description L 8 mm, bar diam. 1.7 mm | description L 35 mm, bar diam. 6 mm |
| manufacturer/tradename BRAND 137109 | manufacturer/tradename BRAND 137114 | manufacturer/tradename BRAND 137107 | manufacturer/tradename BRAND 137127 |
| material PTFE-coated , cylindrical | material PTFE-coated , cylindrical | material PTFE-coated , cylindrical | material PTFE-coated , cylindrical |
| L 13 mm | L 15 mm | L 8 mm | L 35 mm |
| bar diam. 3 mm | bar diam. 6 mm | bar diam. 1.7 mm | bar diam. 6 mm |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
Still not finding the right product?
Explore all of our products under BRAND® magnetic stirring bar, PTFE, cylindrical
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?