Merck
Z328472
BRAND® Transferpettor caps
maximum volume 10 μL
Synonim(y):
glass caps for Transferpettor, glass caps for fixed volume pipettes
Informacje o tej pozycji
NACRES:
NB.01
UNSPSC Code:
41121600
Volume:
10 μL
Sterility:
non-sterile
Feature:
color-coded orange, DE-M marking
Packaging:
pack of 100 ea
Manufacturer/tradename:
BRAND 701902, BRAND
Pomoc techniczna
Pozwól nam pomóc
material
glass
Quality Segment
sterility
non-sterile
feature
color-coded orange, DE-M marking
packaging
pack of 100 ea
manufacturer/tradename
BRAND 701902, BRAND
maximum volume
10 μL
volume
10 μL
Other Notes
BR701902 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Still not finding the right product?
Explore all of our products under BRAND® Transferpettor caps
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)